Carolina™ Blue Box, Electron Microscopy Sciences
Carolina™ Blue Box, Electron Microscopy Sciences
Carolina™ Blue Box, Electron Microscopy Sciences
Certificates
Carolina™ Blue Box, Electron Microscopy Sciences
Carolina™ Blue Box, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Molded from blue high-impact styrene, the Carolina Slide Box is manufactured to exacting specifications.
Compared to conventional wooden boxes, the Carolina Blue Box is smaller in size, lighter in weight, and held to closer dimensional tolerances. The 75 x 25 mm (3 x 1") slides fit the slots and will not slip out once in place. A shoulder on each box excludes dust and identities the top of the box. The boxes are available in two sizes: 3 1/4 x 4 1/4 x 1 3/16" (for 12 slides).
Product Family Options
Product Information
  • Désignation
    Capacité
    Dimensions
    Availability
    Price
  • Carolina Blue Box
    12 Slide
    3.25" x 4.25" x 1.1875"
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $140.43
    $140.43
    Log in for availability
    Pack of 10
    Pack of 10 - $124.20
    $124.20
    Each
       $140.43
    Pack of 10
       $124.20
    Supplier Number: 71482-10
  • Carolina Blue Box
    25 Slide
    Discontinued
    Supplier Number: 71483-01
  • Carolina Blue Box
    25 Slide
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $183.19
    $183.19
    Log in for availability
    Pack of 10
    Pack of 10 - $146.92
    $146.92
    Each
       $183.19
    Pack of 10
       $146.92
    Supplier Number: 71483-10