Nunclon™ Multi-Dishes Plates, Electron Microscopy Sciences | Avantor
Nunclon™ Multi-Dishes Plates, Electron Microscopy Sciences
Nunclon™ Multi-Dishes Plates, Electron Microscopy Sciences
Certificates
Nunclon™ Multi-Dishes Plates, Electron Microscopy Sciences
Nunclon™ Multi-Dishes Plates, Electron Microscopy Sciences
7 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Useful in all areas of cell culture including scale-up and cloning.
  • Raised well rims to lower risk of cross contamination
  • Excellent optical quality
  • Nunclon™ Delta certified
  • Multidish 6 is available coated with Poly-D-Lysine or Collagen
Product Family Options
Informations sur le produit
  • Désignation
    Nb de puits
    Sterile
    Sterility
    Well Shape
    Availability
    Price
  • MultiDish Nunclon Delta
    4
    Sterile
    Sterile
    Round
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 120
    Case of 120 - $1,393.31
    $1,393.31
    Log in pour la disponibilité
    Each
    Each - $1,393.31
    $1,393.31
    Case of 120
       $1,393.31
    Each
       $1,393.31
    Supplier Number: 70426
  • MultiDish
    4
    Sterile
    Sterile
    Round
    Discontinued
    Supplier Number: 70425
  • MultiDish
    4
    Sterile
    Sterile
    Rectangle
    Discontinued
    Supplier Number: 70440
  • MultiDish
    4
    Sterile
    Sterile
    Rectangle
    Discontinued
    Supplier Number: 70440-100
  • MultiDish
    6
    Sterile
    Sterile
    Round
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 8
    Case of 8 - $957.84
    $957.84
    Log in pour la disponibilité
    Each
    Each - $957.84
    $957.84
    Case of 8
       $957.84
    Each
       $957.84
    Supplier Number: 70429
  • MultiDish
    6
    Sterile
    Sterile
    Round
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Bag of 15
    Bag of 15 - $294.29
    $294.29
    Log in pour la disponibilité
    Each
    Each - $294.29
    $294.29
    Bag of 15
       $294.29
    Each
       $294.29
    Supplier Number: 70428
  • MultiDish Collagen Coated
    6
    Non sterile
    Round
    Discontinued
    Supplier Number: 70441
  • MultiDish Nunclon Delta
    6
    Sterile
    Sterile
    Round
    Discontinued
    Supplier Number: 70442
  • MultiDish Poly-D-Lysine Coated
    6
    Non sterile
    Round
    Discontinued
    Supplier Number: 70444
  • MultiDish Collagen Coated
    6
    Non sterile
    Round
    Discontinued
    Supplier Number: 70441-20
  • MultiDish Nunclon Delta
    6
    Sterile
    Sterile
    Round
    Discontinued
    Supplier Number: 70442-75
  • MultiDish Poly-D-Lysine Coated
    6
    Non sterile
    Round
    Discontinued
    Supplier Number: 70444-20
  • MultiDish
    8
    Sterile
    Sterile
    Square
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $239.36
    $239.36
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $180.93
    $180.93
    Each
       $239.36
    Pack of 10
       $180.93
    Supplier Number: 70443
  • MultiDish
    8
    Sterile
    Sterile
    Square
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $2,057.72
    $2,057.72
    Log in pour la disponibilité
    Each
    Each - $2,057.72
    $2,057.72
    Case of 100
       $2,057.72
    Each
       $2,057.72
    Supplier Number: 70443-100
  • MultiDish Nunclon Delta
    12
    Sterile
    Sterile
    Round
    Discontinued
    Supplier Number: 70424
  • MultiDish
    24
    Sterile
    Sterile
    Round
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $32.67
    $32.67
    Each
       $32.67
    Supplier Number: 70448
  • MultiDish
    24
    Sterile
    Sterile
    Round
    Discontinued
    Supplier Number: 70449
  • MultiDish
    24
    Sterile
    Sterile
    Round
    Discontinued
    Supplier Number: 70448-75
  • MultiDish
    24
    Sterile
    Sterile
    Round
    Discontinued
    Supplier Number: 70449-85
  • MultiDish
    48
    Sterile
    Sterile
    Round
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,355.43
    $1,355.43
    Log in pour la disponibilité
    Pack of 48
    Pack of 48 - $1,028.41
    $1,028.41
    Each
       $1,355.43
    Pack of 48
       $1,028.41
    Supplier Number: 70427
  • MultiDish
    48
    Sterile
    Sterile
    Round
    Discontinued
    Supplier Number: 70450
  • MultiDish
    48
    Sterile
    Sterile
    Round
    Discontinued
    Supplier Number: 70450-85