Nunclon Multi-Dishes, Electron Microscopy Sciences | Avantor
Nunclon Multi-Dishes, Electron Microscopy Sciences
Nunclon Multi-Dishes, Electron Microscopy Sciences
Certificates
Nunclon Multi-Dishes, Electron Microscopy Sciences
Nunclon Multi-Dishes, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The NUNC™ Coverglass Optical Bottom 96 Well Plates have an optically clear polystyrene plate bottom. This clear base permits microscopic viewing and reading of well The plate’s upper structure is offered in black for fluorometric detection. External dimension: 128 x 86 mm.
  • White or black upper structure with Polymer bottom combines the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications
  • Optical clarity for viewing well contents
  • Footprint compatible with standard equipment and automated systems
  • Flat bottom well geometry for plate reader access
  • Cell culture, poly-D-Lysine or Collagen I surfaces for cell-based array
  • Non-treated plates are optimized for scintillation counting
  • Working ranges: 50 –200 µl/well
Polystyrene lids have external dimensions of 128 x 86 mm.
Product Family Options
Informations sur le produit
  • Désignation
    Well Volume
    Well Depth
    Color
    Sterility
    Surface Treatment
    Well Diameter
    Availability
    Price
  • 96 well Plate with Lid
    400 µl
    12.2 mm
    Black
    Sterile
    Cell culture
    6.5 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $1,203.98
    $1,203.98
    Log in pour la disponibilité
    Each
    Each - $1,203.98
    $1,203.98
    Case of 30
       $1,203.98
    Each
       $1,203.98
    Supplier Number: 64810-15
  • 96 well Plate with Lid
    400 µl
    12.2 mm
    White
    Sterile
    Cell culture
    6.5 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $1,011.56
    $1,011.56
    Log in pour la disponibilité
    Each
    Each - $1,011.56
    $1,011.56
    Case of 30
       $1,011.56
    Each
       $1,011.56
    Supplier Number: 64810-05
  • 96 well Plate without Lid
    400 µl
    12.2 mm
    Black
    Non sterile
    Untreated
    6.5 mm
    Discontinued
    Supplier Number: 64811-10
  • 96 well Plate without Lid
    400 µl
    12.2 mm
    White
    Non sterile
    Untreated
    6.5 mm
    Discontinued
    Supplier Number: 64811-05
  • 96 well Plate with Lid
    400 µl
    12.2 mm
    Black
    Non sterile
    Collagen I
    6.5 mm
    Discontinued
    Supplier Number: 64810-16
  • 96 well Plate with Lid
    400 µl
    12.2 mm
    White
    Non sterile
    Collagen I
    6.5 mm
    Discontinued
    Supplier Number: 64810-17
  • 96 well Plate with Lid
    400 µl
    12.2 mm
    White
    Non sterile
    Poly-D-Lysine
    6.5 mm
    Discontinued
    Supplier Number: 64810-18
  • 96 well Plate with Lid
    400 µl
    12.2 mm
    Black
    Non sterile
    Poly-D-Lysine
    6.5 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 20
    Case of 20 - $1,746.24
    $1,746.24
    Log in pour la disponibilité
    Each
    Each - $1,746.24
    $1,746.24
    Case of 20
       $1,746.24
    Each
       $1,746.24
    Supplier Number: 64810-19