Flat Mounting Blocks , Electron Microscopy Sciences
Flat Mounting Blocks , Electron Microscopy Sciences
Flat Mounting Blocks , Electron Microscopy Sciences
Certificates
Flat Mounting Blocks , Electron Microscopy Sciences
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Stainless steel flat mounting blocks with the diameter of 25 mm (1") are used for lapping and polishing fixtures.
It is used for the models 150, 151 and 155.
Product Family Options
Informations sur le produit
  • Désignation
    Ø
    Availability
    Price
  • Flat Mounting Blocks (Stainless Steel)
    0.1"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $166.21
    $166.21
    Each
       $166.21
    Supplier Number: 50152-02
  • Flat Mounting Blocks (Stainless Steel)
    0.75"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $145.61
    $145.61
    Each
       $145.61
    Supplier Number: 50152-01
  • Flat Mounting Blocks (Stainless Steel)
    1.25"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $169.24
    $169.24
    Each
       $169.24
    Supplier Number: 50152-03
  • Flat Mounting Blocks (Stainless Steel)
    1.5"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $166.23
    $166.23
    Each
       $166.23
    Supplier Number: 50152-04
  • Flat Mounting Blocks (Stainless Steel)
    2"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $192.31
    $192.31
    Each
       $192.31
    Supplier Number: 50152-05