MicroPoint™ Micro Scissors, Electron Microscopy Sciences
MicroPoint™ Micro Scissors, Electron Microscopy Sciences
MicroPoint™ Micro Scissors, Electron Microscopy Sciences
Certificates
MicroPoint™ Micro Scissors, Electron Microscopy Sciences
MicroPoint™ Micro Scissors, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Featherlite scissors in different styles.
Style DS-Straight Vannas Scissors, 5 mm and smooth blades. Length 125 mm (5").
Style DS-G – Stork Style, 10 mm long and smooth blades. Length: 125 mm (5").
Style DS-GM – Micro Stork Style, 5 mm long and smooth blades. Length: 125 mm (5").
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Featherlite Micro Stork Style Scissors
    Discontinued
    Supplier Number: 77902
  • Featherlite Vannas Scissors Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $772.64
    $772.64
    Each
       $772.64
    Supplier Number: 77900-S
  • Featherlite Stork Style Scissors, Universal
    Discontinued
    Supplier Number: 77901
  • Featherlite Vannas Scissors Curved
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $667.12
    $667.12
    Each
       $667.12
    Supplier Number: 77900-C