Formaldehyde Spill Response™ Kit, Electron Microscopy Sciences
Product Details & Documents
This kit features FSC-1™ to aid in reducing exposure to formaldehyde gas in the event of a spill. The formalin is transformed into an easily removable semi solid mass. The resulting mass cools rapidly to minimize formaldehyde off gassing. Kits contains 1 – 11oz bottle of FSC-1™ solidifier, 1 pair of vented goggles, a respirator, an open backed gown to protect clothing, a pair of nitrile gloves, 4 scoops and scrapers, 3 wiper pads and 4 thirty gallon bags.
Spécifications
- Désignation:Formaldehyde Spill Response Refill