Top-Loading Dispensing Bins, Electron Microscopy Sciences
Top-Loading Dispensing Bins, Electron Microscopy Sciences
Top-Loading Dispensing Bins, Electron Microscopy Sciences
Certificates
Top-Loading Dispensing Bins, Electron Microscopy Sciences
Top-Loading Dispensing Bins, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Bins are made from ¹/₄" thick PVC. They are chemical resistant and easy to clean.
  • Removable top for easy loading
  • Hinged clear acrylic door
  • Different sizes for your needs
Ideal for pipette tips, small vials, cassette lids, cassette base molds, cotton swabs, aluminum stubs, embedding capsules etc.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Medium Top-Loading Dispensing Bins
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 391,73 $
    391,73 $
    Each
       391,73 $
    Supplier Number: 64388-12
  • Large Top-Loading Dispensing Bins
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 462,59 $
    462,59 $
    Each
       462,59 $
    Supplier Number: 64388-18
  • Small Top-Loading Dispensing Bins
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 298,73 $
    298,73 $
    Each
       298,73 $
    Supplier Number: 64388-05