Pipettor Holder, Electron Microscopy Sciences | Avantor
Pipettor Holder, Electron Microscopy Sciences
Pipettor Holder, Electron Microscopy Sciences
Certificates
Pipettor Holder, Electron Microscopy Sciences
Pipettor Holder, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from High Density Polyethylene. Accommodate most major brands of pipettors.
  • Resistant to most solvents and is easy to clean
  • Supplied with non-skid feet
  • Available in 4-place and 6-place versions
Also Available in Black HDPE/UV, perfect for use in DNA Workstations, where U.V. anti-germicidal amp are used.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • 4 Place Pipettor Holder
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 229,38 $
    229,38 $
    Each
       229,38 $
    Supplier Number: 64378-04
  • 6 Place Pipettor Holder
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 275,13 $
    275,13 $
    Each
       275,13 $
    Supplier Number: 64378-06