Coverplate Microwave Immunostaining Systems, Electron Microscopy Sciences
Coverplate Microwave Immunostaining Systems, Electron Microscopy Sciences
Coverplate Microwave Immunostaining Systems, Electron Microscopy Sciences
The Coverplate Microwave Immunostaining Systems. Shandon Lipshaw's patented Coverplate technology is the foundation of our microwave immunostaining system. Increased quality control, time and reagent savings, specimen protection, and consistent superb quality stains can be expected when using our Coverplate Microwave Immunostaining protocols.
Product Family Options
  • Désignation
  • Disposable Cover Plates
    Each
    Each - $137.05
    $137.05
    Pack of 10
    Pack of 10 - $121.36
    $121.36
    Each
       $137.05
    Pack of 10
       $121.36
     : 97091
  • Cover Plate Slide Racks
    Each
    Each - $1,455.58
    $1,455.58
    Pack of 1
    Pack of 1 - $1,155.28
    $1,155.28
    Each
       $1,455.58
    Pack of 1
       $1,155.28
     : 97092