ContactUsLinkComponent
Cassette Racks for Microwave Histoprocessing, Electron Microscopy Sciences
Catalog # 100492-520
Minority or Woman-Owned Business Enterprise
Cassette Racks for Microwave Histoprocessing, Electron Microscopy Sciences
Catalog # 100492-520
Supplier Number:  97050
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    PTFE Cassette Rack for Microwave Histoprocessing
  • Cat. no.:
    100492-520
  • Supplier No.:
    97050
Specifications
About this item
Simple and reliable device designed to hold up to 24 standard cassettes for histoprocessing in a microwave. This new rack is manufactured from "PTFE" resins, and will withstand repeated exposure to fixatives, dehydrating agents, clearing agents and paraffin. It is easily cleaned in hot water. It is intended for use with a disposable tray which holds the processing chemicals.
Four processing racks and trays will fit into the EMS 9000 Microwave Processor, so that up to 96 cassettes can be processed in one run. Using this system, the histotechnologist needs to handle the individual cassettes only once, to load them into the racks. The rack is then placed into a tray containing the dehydrating medium, usually 100% ethyl alcohol, and microwaved. After dehydration, the rack containing the cassettes is transferred into a tray containing the clearing agent, usually isopropanol, and microwaved. After clearing, the rack is transferred to a tray containing liquid paraffin, and microwaved. The whole process can take as little as 25 minutes (for small specimens, such as needle biopsies). There are no hazardous chemicals needed (no xylene!).