Tungsten Wire Filaments, Electron Microscopy Sciences
À propos de cet article
V-shaped filaments are high purity, vacuum grade tungsten wire 0.02" (0.50 mm) diameter, V-shaped filaments ½" (12.7mm) deep, with a bottom angle of 45°, either single wire or triple strand.
Coiled filaments are single or 3-strand coiled tungsten filaments, 0.5 mm (0.002") diameter wires.
4 Options Available Below
Product Details & Documents
V-shaped filaments are high purity, vacuum grade tungsten wire 0.02" (0.50 mm) diameter, V-shaped filaments ½" (12.7mm) deep, with a bottom angle of 45°, either single wire or triple strand.
Coiled filaments are single or 3-strand coiled tungsten filaments, 0.5 mm (0.002") diameter wires.
Product Family Options
Informations sur le produit
- DésignationØLongueurAvailabilityPrice
- Triple tungsten V-shaped filaments, 5/8" (16 mm) v-opening0.5 mm (0.02")3"Each97,74 $Pack of 1086,46 $Supplier Number: 73842
- Single tungsten V-shaped filaments, 3/4" (19 mm) v-opening0.5 mm (0.02")3"Each46,55 $Pack of 1039,22 $Supplier Number: 73840
- Tungsten coiled filaments, triple strand 12 turns9.5 mm (0.5")5"Each116,77 $Pack of 593,36 $Supplier Number: 73939-1
- Tungsten coiled filaments, single strand 6 turns6.3 mm (0.25")4"Each95,12 $Pack of 571,19 $Supplier Number: 73938-1
- Triple tungsten V-shaped filaments, 3/4" (19 mm) v-opening0.02" (0.50 mm)3.5"Discontinued-Supplier Number: 73844
- Tungsten coiled filaments, triple strand 6 turns6.3 mm (0.25")4"Discontinued-Supplier Number: 73938-3