Tungsten Wire, Electron Microscopy Sciences
Tungsten Wire, Electron Microscopy Sciences
Tungsten Wire, Electron Microscopy Sciences
Tungsten Wire, Electron Microscopy Sciences
We are offering tungsten wire in 99.90% purity.
Available in 0.025" (.64 mm), 0.02" (.51 mm), 0.009" (0.23 mm) diameters.
Product Family Options
  • Désignation
    Ø
    Longueur
  • Tungsten wire
    0.009" (0.25 mm)
    20 ft.
    Each
    Each - $135.79
    $135.79
    Each
       $135.79
     : 73804
  • Tungsten wire
    0.02" (0.51 mm)
    20 ft.
    Each
    Each - $114.74
    $114.74
    Each
       $114.74
     : 73800
  • Tungsten wire
    0.025" (0.64 mm)
    100 ft.
    Each
    Each - $977.51
    $977.51
    Each
       $977.51
     : 73803