Kendall Treated Pipette Tips, Electron Microscopy Sciences | Avantor
Kendall Treated Pipette Tips, Electron Microscopy Sciences
Kendall Treated Pipette Tips, Electron Microscopy Sciences
Certificates
Kendall Treated Pipette Tips, Electron Microscopy Sciences
Kendall Treated Pipette Tips, Electron Microscopy Sciences
14 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
KENDALL’s low binding tips ensure accuracy and precision by bonding a proprietary polymer to the resin. Precision and accuracy are increasingly important in molecular biology labs. A pipetter can be 99% accurate and precise, but the tip can com-promise performance by retaining or binding a sample.
  • Not detectable/Non-reactive
  • Extracted only by a fluorinated solvent
  • Autoclaving will not compromise performance
  • Tips and racks are fully autoclavable
  • Graduation marks at 2 and 5 µl
  • Yellow tips non-graduated, not-treated
  • Available bulk, racked, racked sterile, EcoStack racks and QuickRack tip reloading system
Reduces the surface energy of liquid and repels any substance. Extended length reaches bottom of tubes without contaminating the pipettor shaft. Low Binding Polymer Technology: A manufacturing process that binds a perfluorooctyl polymer to polypropylene resin–resulting in an exposed trifluoromethyl surface.
Graduations:
0.1 to 10 µl: 2 and 5 µl
5 to 200 µl / 5 to 300 µl / 101 to 1000 µl: 10, 50 and 100 µl
Available in convenient hinged racked package, with and without low binding surface. Available bulk (not treated), racked and racked sterile (low binding).
Available in QuickRack
0.5 to 10 µl Crystal/Eppendorf Tips-Natural: Fits Eppendorf ultra micro pipetters
1 to 200 µl Universal Tips: Universal fit for research-grade pipetters
P2/P10 Ultra Micro Tip – Natural: Universal fit for ultra-micro pipetters
Product Family Options
P2/P10 Ultra Micro Tips
Informations sur le produit
  • Tip Style
    Volume
    Sterile
    Tip Color
    Packed
    Availability
    Price
  • P2 Ultra Micro Tips, graduated
    0.1 - 10 µl
    Naturel
    1000/Bag, Bulk Packed
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Bag of 1,000
    Bag of 1,000 - $96.86
    $96.86
    Log in pour la disponibilité
    Each
    Each - $96.86
    $96.86
    Bag of 1,000
       $96.86
    Each
       $96.86
    Supplier Number: 72440-01
  • P2 Ultra Micro Tips, graduated
    0.1 - 10 µl
    Naturel
    2 Racks/Case, 96 Tips/Rack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 200
    Case of 200 - $72.41
    $72.41
    Log in pour la disponibilité
    Each
    Each - $72.41
    $72.41
    Case of 200
       $72.41
    Each
       $72.41
    Supplier Number: 72441-01
  • P2 Ultra Micro Tips, graduated
    0.1 - 10 µl
    +
    Naturel
    2 Racks/Case, 96 Tips/Rack
    Discontinued
    Supplier Number: 72441-02
  • P2 Ultra Micro Tips, Low Retention, graduated
    0.1 - 10 µl
    Naturel
    10 Racks/Case, 96 Tips/Rack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $198.64
    $198.64
    Log in pour la disponibilité
    Each
    Each - $198.64
    $198.64
    Case of 10
       $198.64
    Each
       $198.64
    Supplier Number: 72440-02
  • P2 Ultra Micro Tips, Low Retention, graduated
    0.1 - 10 µl
    +
    Naturel
    10 Racks/Case, 96 Tips/Rack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $183.10
    $183.10
    Log in pour la disponibilité
    Each
    Each - $183.10
    $183.10
    Case of 10
       $183.10
    Each
       $183.10
    Supplier Number: 72440-03
  • P2 Ultra Micro Tips, Low Retention, graduated
    0.1 - 10 µl
    +
    Naturel
    10 Racks/Case, 96 Tips/Quick Rack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $343.98
    $343.98
    Log in pour la disponibilité
    Each
    Each - $343.98
    $343.98
    Case of 10
       $343.98
    Each
       $343.98
    Supplier Number: 72440-04
Crystal/Eppendorf Tips
Informations sur le produit
  • Tip Style
    Volume
    Sterile
    Tip Color
    Packed
    Availability
    Price
  • Crystal Eppendorf Tips, graduated
    0.1 - 10 µl
    Naturel
    1000/Bag, Bulk Packed
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Bag of 1,000
    Bag of 1,000 - $299.82
    $299.82
    Log in pour la disponibilité
    Each
    Each - $299.82
    $299.82
    Bag of 1,000
       $299.82
    Each
       $299.82
    Supplier Number: 72442-01
  • Crystal Eppendorf Tips, Low Retention, graduated
    0.1 - 10 µl
    Naturel
    10 Racks/Case, 96 Tips/Rack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 960
    Case of 960 - $141.71
    $141.71
    Log in pour la disponibilité
    Each
    Each - $141.71
    $141.71
    Case of 960
       $141.71
    Each
       $141.71
    Supplier Number: 72442-02
Universal Tips
Informations sur le produit
  • Tip Style
    Volume
    Sterile
    Tip Color
    Packed
    Availability
    Price
  • Universal Tips, graduated
    5 - 200 µl
    Naturel
    1000/Bag, Bulk Packed
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Bag of 1,000
    Bag of 1,000 - $61.73
    $61.73
    Log in pour la disponibilité
    Each
    Each - $61.73
    $61.73
    Bag of 1,000
       $61.73
    Each
       $61.73
    Supplier Number: 72443-01
  • Universal Tips, graduated
    5 - 200 µl
    Naturel
    1000/Bag, Bulk Packed
    Discontinued
    Supplier Number: 72444-01
  • Universal Tips, graduated
    101 - 1000 µL
    Naturel
    1000/Bag, Bulk Packed
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Bag of 1,000
    Bag of 1,000 - $86.03
    $86.03
    Log in pour la disponibilité
    Each
    Each - $86.03
    $86.03
    Bag of 1,000
       $86.03
    Each
       $86.03
    Supplier Number: 72445-01
  • Universal Tips, Low Retention, graduated
    5 - 200 µl
    Naturel
    10 Racks/Case, 96 Tips/Rack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $131.78
    $131.78
    Log in pour la disponibilité
    Each
    Each - $131.78
    $131.78
    Case of 10
       $131.78
    Each
       $131.78
    Supplier Number: 72443-02
  • Universal Tips, Low Retention, graduated
    5 - 200 µl
    +
    Naturel
    10 Racks/Case, 96 Tips/Rack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 960
    Case of 960 - $148.62
    $148.62
    Log in pour la disponibilité
    Each
    Each - $148.62
    $148.62
    Case of 960
       $148.62
    Each
       $148.62
    Supplier Number: 72443-03
  • Universal Tips, Low Retention, graduated
    5 - 200 µl
    +
    Naturel
    10 Racks/Case, 96 Tips/Eco Rack
    Discontinued
    Supplier Number: 72443-04
  • Universal Tips, Low Retention, graduated
    5 - 200 µl
    Naturel
    10 Racks/Case, 96 Tips/Quick Rack
    Discontinued
    Supplier Number: 72443-05
  • Universal Tips, Low Retention, graduated
    5 - 300 µl
    +
    Naturel
    10 Racks/Case, 96 Tips/Rack
    Discontinued
    Supplier Number: 72444-03
  • Universal Tips, Low Retention, graduated
    101 - 1000 µL
    Naturel
    10 Racks/Case, 96 Tips/Rack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $91.44
    $91.44
    Log in pour la disponibilité
    Each
    Each - $91.44
    $91.44
    Case of 10
       $91.44
    Each
       $91.44
    Supplier Number: 72445-02
  • Universal Tips, Low Retention, graduated
    101 - 1000 µL
    +
    Naturel
    10 Racks/Case, 96 Tips/Rack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $156.96
    $156.96
    Log in pour la disponibilité
    Each
    Each - $156.96
    $156.96
    Case of 10
       $156.96
    Each
       $156.96
    Supplier Number: 72445-03
  • Universal Tips, Not Treated
    5 - 200 µl
    Jaune
    1000/Bag, Bulk Packed
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Bag of 1,000
    Bag of 1,000 - $76.85
    $76.85
    Log in pour la disponibilité
    Each
    Each - $76.85
    $76.85
    Bag of 1,000
       $76.85
    Each
       $76.85
    Supplier Number: 72446-01
  • Universal Tips, Not Treated
    5 - 200 µl
    Jaune
    10 Racks/Case, 96 Tips/Rack
    Discontinued
    Supplier Number: 72446-02
  • Universal Tips, Not Treated
    5 - 200 µl
    +
    Jaune
    10 Racks/Case, 96 Tips/Rack
    Discontinued
    Supplier Number: 72446-03