High Opacity Paint Markers, Electron Microscopy Sciences
High Opacity Paint Markers, Electron Microscopy Sciences
High Opacity Paint Markers, Electron Microscopy Sciences
Certificates
High Opacity Paint Markers, Electron Microscopy Sciences
High Opacity Paint Markers, Electron Microscopy Sciences
6 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Constructed aluminum barrel, actuating ball and hard felt tips, these markers permanently cover nearly any surface
  • Weatherproof
  • Resistant to alcohol and acetone
  • Contain xylene
Markers are available in black, red and white with bullet tips. Except model 130, it writes like a pen with white ink for marking on dark surfaces, including carbon tabs, which is use in SEM specimen mounting purpose.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Large Marker, Black
    Log in for availability
    Log in for availability
    Each
    Each - $16.86
    $16.86
    Each
       $16.86
    Supplier Number: 72169-B2
  • Large Marker, White
    Log in for availability
    Log in for availability
    Each
    Each - $16.86
    $16.86
    Each
       $16.86
    Supplier Number: 72168-W2
  • Opaque White Pen
    Log in for availability
    Log in for availability
    Each
    Each - $19.17
    $19.17
    Each
       $19.17
    Supplier Number: 72168-W1
  • Small Marker, Black
    Log in for availability
    Log in for availability
    Each
    Each - $9.14
    $9.14
    Each
       $9.14
    Supplier Number: 72168-B1
  • Small Marker, Red
    Log in for availability
    Log in for availability
    Each
    Each - $9.04
    $9.04
    Each
       $9.04
    Supplier Number: 72167-R1
  • Large Marker, Red
    Log in for availability
    Log in for availability
    Each
    Each - $16.62
    $16.62
    Each
       $16.62
    Supplier Number: 72167-R2