About this item
Design minimize friction, promotes reagent mixing, and facilitates uniform hybridization.
- Sealable access ports (1.5 mm) in the chamber surface allow for the addition and removal of reactants
- Adhesive seal tabs create leak-proof chambers that are temperature resistant
- SecureSeal™adhesive bonds chambers to glass in seconds and removes cleanly and easily even after heating
- SecureSeals™are ideally suited for protocols utilizing autoradiographic, fluorescent or cheminescent end points
- Polycarbonate HybriSlip used, 0.25 mm thick
SecureSeal™ adhesive chambers form peel-and-stick enclosures to isolate specimens affixed to glass microscope slides. RNase free, gasketed chambers provide optimum surface-to-volume fluid dynamics for hybridization assays on large or multiple specimens and genomic arrays on glass slides or film coated slides.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.