Polystyrene Containers, Electron Microscopy Sciences
Polystyrene Containers, Electron Microscopy Sciences
Polystyrene Containers, Electron Microscopy Sciences
Certificates
Polystyrene Containers, Electron Microscopy Sciences
Polystyrene Containers, Electron Microscopy Sciences
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These clear and rigid containers are made from polystyrene.
  • All containers are aseptically manufactured
  • Labeling options include unlabeled, printed, or plain
  • 30mL containers with universal closure rely on special seal technology
  • Other sizes employ liners or crab claw closures to facilitate sample containment
  • Some sizes are available with spoons
  • For in vitro use only
Product Family Options
Product Information
  • Capacité
    Désignation
    Hauteur
    Cap Type
    Label Type
    Ø
    Availability
    Price
  • 60 ml
    Container
    61.1 mm
    PE, universal cap
    Plain
    4.6 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $416.19
    $416.19
    Log in for availability
    Pack of 300
    Pack of 300 - $317.41
    $317.41
    Each
       $416.19
    Pack of 300
       $317.41
    Supplier Number: 64240-01
  • 30 ml
    Container with Spoon
    93.1 mm
    PP, universal cap
    Plain
    30.1 mm
    Discontinued
    Supplier Number: -
  • 150 ml
    Container
    107.5 mm
    Meta cap
    Plain
    54.9 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $1,014.80
    $1,014.80
    Log in for availability
    Pack of 120
    Pack of 120 - $794.08
    $794.08
    Each
       $1,014.80
    Pack of 120
       $794.08
    Supplier Number: 64240-09
  • 60 ml
    Container
    61.1 mm
    PE, plain cap
    Printed
    44.6 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $597.01
    $597.01
    Log in for availability
    Pack of 300
    Pack of 300 - $455.10
    $455.10
    Each
       $597.01
    Pack of 300
       $455.10
    Supplier Number: 64240-13
  • 30 ml
    Container
    93.1 mm
    PP, universal cap
    Printed
    30.1 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $1,346.48
    $1,346.48
    Log in for availability
    Pack of 400
    Pack of 400 - $1,026.94
    $1,026.94
    Each
       $1,346.48
    Pack of 400
       $1,026.94
    Supplier Number: 64240-03
  • 30 ml
    Container with Spoon
    93.1 mm
    PE, universal cap
    Printed
    30.1 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $897.16
    $897.16
    Log in for availability
    Pack of 400
    Pack of 400 - $746.85
    $746.85
    Each
       $897.16
    Pack of 400
       $746.85
    Supplier Number: 64240-11
  • Discontinued
    Supplier Number: -