DrainRack™ Jr. Slide Rack, Electron Microscopy Sciences | Avantor
DrainRack™ Jr. Slide Rack, Electron Microscopy Sciences
DrainRack™ Jr. Slide Rack, Electron Microscopy Sciences
Certificates
DrainRack™ Jr. Slide Rack, Electron Microscopy Sciences
DrainRack™ Jr. Slide Rack, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from high impact polystyrene. This model is similar to the standard DrainRack™ but can hold up to 100 microscope slides in 50 individual numbered slots.
The rack is autoclavable. Dimensions: 75 x 125 x 25 mm (3 x 5 x 1").
Product Family Options
Informations sur le produit
  • Color
    Availability
    Price
  • Green
    Discontinued
    Supplier Number: 71445-G
  • Pink
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $83.80
    $83.80
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $66.12
    $66.12
    Each
       $83.80
    Pack of 10
       $66.12
    Supplier Number: 71445-P
  • White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $83.80
    $83.80
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $66.12
    $66.12
    Each
       $83.80
    Pack of 10
       $66.12
    Supplier Number: 71445-W
  • Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $84.94
    $84.94
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $77.06
    $77.06
    Each
       $84.94
    Pack of 10
       $77.06
    Supplier Number: 71445-B
  • Yellow
    Discontinued
    Supplier Number: 71445-Y