Process Embedding Cassettes, Electron Microscopy Sciences
À propos de cet article
Made from Acetal copolymer – Disposable plastic tissue cassettes are suitable for holding and identifying tissue samples in processing, embedding, and sectioning procedures. The cassettes fit securely in microtome chuck adapters. They are molded from a high-density polymer that is totally resistant to histological solvents. These cassettes are designed to accept standard metal lids and will also fit either the standard stainless steel base molds or the disposable plastic ones. Only Clear cassettes are made of polypropylene. All other cassettes are made of Acetal Polymer. Each case contains 3 dispenser boxes of 500 cassettes. Stainless Steel Lids order separately.
10 Options Available Below
Product Details & Documents
Made from Acetal copolymer – Disposable plastic tissue cassettes are suitable for holding and identifying tissue samples in processing, embedding, and sectioning procedures. The cassettes fit securely in microtome chuck adapters. They are molded from a high-density polymer that is totally resistant to histological solvents. These cassettes are designed to accept standard metal lids and will also fit either the standard stainless steel base molds or the disposable plastic ones. Only Clear cassettes are made of polypropylene. All other cassettes are made of Acetal Polymer. Each case contains 3 dispenser boxes of 500 cassettes. Stainless Steel Lids order separately.
Product Family Options
Informations sur le produit
- DésignationColorAvailabilityPrice
- Process/Embedding CassettesBlueDiscontinued-Supplier Number: 70025-B
- Process/Embedding Cassettes w/o LidsLilacCase of 500$150.64Each$150.64Supplier Number: 70025-L
- Process/Embedding Cassettes w/o LidOrangeCase of 500$150.64Each$150.64Supplier Number: 70025-O
- Process/Embedding Cassettes w/o LidsTanCase of 500$150.64Each$150.64Supplier Number: 70025-T
- Process/Embedding Cassettes w/o LidsClearCase of 500$150.64Each$150.64Supplier Number: 70025-C
- Process/Embedding CassettesYellowCase of 1,000$170.36Each$170.36Supplier Number: 70025-Y
- Process/Embedding Cassettes w/o LidsGrayCase of 500$126.89Each$126.89Supplier Number: 70025-GR
- Stainless Steel CoversEach$792.23Pack of 100$604.67Supplier Number: 70025-11
- Process/Embedding CassettesPinkCase of 500$150.64Each$150.64Supplier Number: 70025-P
- Process/Embedding Cassettes w/o LidsPeachCase of 500$150.64Each$150.64Supplier Number: 70025-PE
- Stainless Steel CoversEach$218.14Pack of 25$166.52Supplier Number: 70025-10