Biopsy Processing Embedding Cassettes, Electron Microscopy Sciences
À propos de cet article
Biopsy processing cassettes are similar to the slotted cassettes but have a 1 mm square mesh for small biopsies. Made from Acetyl Polymer, which is a special high density polymer that keeps specimens safely submerged in solution, these cassettes are totally resistant to most histological solvents and their slots offer maximum flow through for good fluid exchange and proper drainage.
- Lids either come attached to the cassette or are packaged separately
- Lids can be opened and closed as often as necessary and always relock securely without specimen loss
- Large labeling areas are located on three sides of the cassette for your convenience
- Each slot measures: 1 x 5 mm, total 128 slots per cassette
Packaging: Cassettes with lids are placed on cassettes, each case contains 3 dispenser boxes of 500 cassettes with lids already locked in place. Cassettes with lids packed separately, each case contains two dispenser boxes of 500 cassettes and Lids.
7 Options Available Below
Product Details & Documents
Biopsy processing cassettes are similar to the slotted cassettes but have a 1 mm square mesh for small biopsies. Made from Acetyl Polymer, which is a special high density polymer that keeps specimens safely submerged in solution, these cassettes are totally resistant to most histological solvents and their slots offer maximum flow through for good fluid exchange and proper drainage.
- Lids either come attached to the cassette or are packaged separately
- Lids can be opened and closed as often as necessary and always relock securely without specimen loss
- Large labeling areas are located on three sides of the cassette for your convenience
- Each slot measures: 1 x 5 mm, total 128 slots per cassette
Packaging: Cassettes with lids are placed on cassettes, each case contains 3 dispenser boxes of 500 cassettes with lids already locked in place. Cassettes with lids packed separately, each case contains two dispenser boxes of 500 cassettes and Lids.
Product Family Options
Informations sur le produit
- DésignationColorUnitsAvailabilityPrice
- Square Mesh Cassettes Lids Packed SeparateAqua500Case of 500$462.59Each$462.59Supplier Number: 70072-A
- Square Mesh Cassettes Lids Packed SeparateBlue—Case of 1,000$462.59Each$462.59Supplier Number: 70072-B
- Square Mesh Cassettes Lids Packed SeparateGray500Discontinued-Supplier Number: 70072-GR
- Square Mesh Cassettes Lids Packed SeparateLilac500Discontinued-Supplier Number: 70072-L
- Square Mesh Cassettes Lids Packed SeparatePink—Case of 1,000$462.59Each$462.59Supplier Number: 70072-P
- Square Mesh Cassettes Lids Packed SeparatePeach500Discontinued-Supplier Number: 70072-PE
- Square Mesh Cassettes with Lids AttachedAqua—Discontinued-Supplier Number: 70073-A
- Square Mesh Cassettes with Lids AttachedBlue—Case of 1,000$703.18Each$703.18Supplier Number: 70073-B
- Square Mesh Cassettes with Lids AttachedGreen—Discontinued-Supplier Number: 70073-G
- Square Mesh Cassettes with Lids AttachedLilac—Discontinued-Supplier Number: 70073-L
- Square Mesh Cassettes with Lids AttachedOrange—Case of 500$652.94Each$652.94Supplier Number: 70073-O
- Square Mesh Cassettes with Lids AttachedPeach—Discontinued-Supplier Number: 70073-PE
- Square Mesh Cassettes with Lids AttachedWhite—Discontinued-Supplier Number: 70073-W
- Square Mesh Cassettes with Lids AttachedYellow—Case of 1,000$703.18Each$703.18Supplier Number: 70073-Y
- Square Mesh Cassettes, Lids Packed Separate, TanTan500Discontinued-Supplier Number: 70072-T
- Square Mesh Cassettes, White, Lids SepWhite—Case of 1,000$498.16Each$498.16Supplier Number: 70072-W