Slide Wing Handle Rack, 30, Electron Microscopy Sciences | Avantor
Slide Wing Handle Rack, 30, Electron Microscopy Sciences
30 Slide Wing Handle Rack
Catalog # 100492-940
Fournisseur #:  70312-34
Slide Wing Handle Rack, 30, Electron Microscopy Sciences
Catalog # 100492-940
Slide Wing Handle Rack, 30, Electron Microscopy Sciences
Slide Wing Handle Rack, 30, Electron Microscopy Sciences
30 Slide Wing Handle Rack
Price
$241.97
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This 30-slide rack is made from non-tarnishing stainless steel and it is resistant to most staining solutions.
The handle is permanently attached but it is swing-able to permitting for closure of the dish and easy insertion and removal of the slides.
Spécifications
  • Désignation:
    30 Slide Wing Handle Rack