Slide Rack, ²⁰/₄₀, Electron Microscopy Sciences
20/40 Slide Wing Handle Rack
  100492-944
 :  70312-36
Slide Rack, ²⁰/₄₀, Electron Microscopy Sciences
  100492-944
Slide Rack, ²⁰/₄₀, Electron Microscopy Sciences
Slide Rack, ²⁰/₄₀, Electron Microscopy Sciences
20/40 Slide Wing Handle Rack
327,03 $
Each
This glass rack holds 20 single slides, or 40 slides back-to-back. Made from soda-lime glass.
  • Désignation:
    20/40 Slide Wing Handle Rack