Glass Staining Dish - 25 Slide Unit, Electron Microscopy Sciences
À propos de cet article
These glass staining dishes are made of a heavy glass with snug fit lids.
- Snug fit lids are included
- Dishes have gasket seal
The stainless steel rack has a swing handle for easy access and has grooves to take 76 mm microscope slides, up to 40 mm wide horizontally.
Inside Dimensions: 33/4 x 3 x 21/4" (H)
Outside Dimension: 41/4 x 35/8 x 3" (H)
: Complete set includes one staining dish with lid and one swing handle rack.
3 Options Available Below
Product Details & Documents
These glass staining dishes are made of a heavy glass with snug fit lids.
- Snug fit lids are included
- Dishes have gasket seal
The stainless steel rack has a swing handle for easy access and has grooves to take 76 mm microscope slides, up to 40 mm wide horizontally.
Inside Dimensions: 33/4 x 3 x 21/4" (H)
Outside Dimension: 41/4 x 35/8 x 3" (H)
: Complete set includes one staining dish with lid and one swing handle rack.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 71420-25
- Supplier Number: 71420-DL
- Supplier Number: 71420-SR