Gold Seal Plain Microscope Slides, Electron Microscopy Sciences
À propos de cet article
GOLDSEAL® slides are made from pure white glass (sodalime) and at accurately cut sizes.
To preserve the highest quality from manufacturer to ultimate user, all GOLD SEAL® slides are hermetically sealed in our Moisture-Barrier™ system, providing a dry-slide environments during shipment and storage.
4 Options Available Below
Product Details & Documents
GOLDSEAL® slides are made from pure white glass (sodalime) and at accurately cut sizes.
To preserve the highest quality from manufacturer to ultimate user, all GOLD SEAL® slides are hermetically sealed in our Moisture-Barrier™ system, providing a dry-slide environments during shipment and storage.
Product Family Options
Informations sur le produit
- DésignationCapacitéÉpaisseurL×lAvailabilityPrice
- Supplier Number: 63710-25
- Cytologenitics1 g1.0 mm12 mmEach$135.88Gross of 144$135.88Supplier Number: 63712-05
- Plain slides1 g1.2 mm75×25 mm (3×1")Each$119.11Gross of 144$119.11Supplier Number: 63711-05
- Plain slides1 g1.2 mm75×25 mm (3×1")Discontinued-Supplier Number: -
- Cytologenitics1 g1.0 mm75×25 mm (3×1")Discontinued-Supplier Number: -
- Plain slides1 g1.0 mm75×25 mm (3×1")Each$128.14Gross of 144$128.14Supplier Number: 63710-05