Specialty Expiration Labels, Electron Microscopy Sciences
Product Details & Documents
Quickly identify outdated laboratory items such as reagents and controls. These labels provide space for noting the date that the items were received, opened and when it will expire.
- The label measures ¾×1½"
A permanent adhesive holds the label firmly in place on a variety of surfaces.
Spécifications
- Désignation:Expiration labels