Microscope Slide Labels, Electron Microscopy Sciences
À propos de cet article
These specialty Microscope Slide Labels and matching End Labels are available in standard (thin) or pathology (tissue high) thickness, and square or round corner (RC).
- Standard thin or tissue high thickness for pathology
- Permanent adhesive holds labels during use
- Can be used for long-term storage
Sheet form size is 5¼×8".
2 Options Available Below
Product Details & Documents
These specialty Microscope Slide Labels and matching End Labels are available in standard (thin) or pathology (tissue high) thickness, and square or round corner (RC).
- Standard thin or tissue high thickness for pathology
- Permanent adhesive holds labels during use
- Can be used for long-term storage
Sheet form size is 5¼×8".
Product Family Options
Informations sur le produit
- DésignationDimensionsÉpaisseurAvailabilityPrice
- Supplier Number: 77022-06
- Microscope slide labels, SLSP 150.94×0.94"PathologyDiscontinued-Supplier Number: 77022-10
- Microscope slide end labels, SESP-160.47×0.94"PathologyDiscontinued-Supplier Number: 77021-55
- Microscope slide labels, SLR-15, roll0.94×0.94"StandardDiscontinued-Supplier Number: 77020-05
- Microscope slide end labels, pathology, roll0.47×0.94"StandardEach$99.75Roll of 1,000$99.75Supplier Number: 77021-05
- Microscope slide labels, SLR15-RC, roll0.94×0.94"StandardDiscontinued-Supplier Number: 77020-06
- Microscope slide labels, SLRP-15, roll0.94×0.94"PathologyDiscontinued-Supplier Number: 77020-10
- Microscope slide labels, SLRP-15-RC, roll0.94×0.94"PathologyDiscontinued-Supplier Number: 77020-11
- Microscope slide end labels, SER-16, roll0.47×0.94"StandardDiscontinued-Supplier Number: 77020-50
- Microscope slide labels, SER-16 rc, roll0.47×0.94"StandardDiscontinued-Supplier Number: 77020-51
- Microscope slide labels, SERP-16, roll0.47×0.94"PathologyDiscontinued-Supplier Number: 77020-55
- Microscope slide labels, SERP-16-RC, roll0.47×0.94"PathologyDiscontinued-Supplier Number: 77020-56