Hi/Lo Write-On Temperature Tape, Electron Microscopy Sciences
À propos de cet article
All rolls fit on a standard 25.4 mm (1") or 76 mm (3") core
These color-coded label tapes work well with all glass and plastic labware, racks, tubing, bags, boxes, and cabinets. It adheres to any surface and peels off without leaving a sticky residue. Tapes resist moisture and most solvents and can withstand autoclaving (250°F). It may be used as well down to -73°C.
All rolls fit on a standard 25.4 mm (1") or 76 mm (3") core. Tape is available in widths of 12.7 mm (1/2"), 19 mm (3/4"), and 25.4 mm (1"). Measures 40 yards (36.6 M) long.
Tape is available in widths of 12.7 mm (1/2"), 19 mm (3/4"), and 25.4 mm (1"). Measures 40 yards (36.6 M) long.
19 Options Available Below
Product Details & Documents
All rolls fit on a standard 25.4 mm (1") or 76 mm (3") core
These color-coded label tapes work well with all glass and plastic labware, racks, tubing, bags, boxes, and cabinets. It adheres to any surface and peels off without leaving a sticky residue. Tapes resist moisture and most solvents and can withstand autoclaving (250°F). It may be used as well down to -73°C.
All rolls fit on a standard 25.4 mm (1") or 76 mm (3") core. Tape is available in widths of 12.7 mm (1/2"), 19 mm (3/4"), and 25.4 mm (1"). Measures 40 yards (36.6 M) long.
Tape is available in widths of 12.7 mm (1/2"), 19 mm (3/4"), and 25.4 mm (1"). Measures 40 yards (36.6 M) long.
Product Family Options
Informations sur le produit
- ColorLongueurLargeurAvailabilityPrice
- White36.6 m (40 yds.)12.7 mm (½")Each41,34 $Roll of 141,34 $Supplier Number: 77010-W1
- Green36.6 m (40 yds.)12.7 mm (½")Each34,25 $Roll of 134,25 $Supplier Number: 77010-G1
- Green36.6 m (40 yds.)19.0 mm (¾")Each41,76 $Roll of 141,76 $Supplier Number: 77010-G2
- Yellow36.6 m (40 yds.)19.0 mm (¾")Discontinued-Supplier Number: 77010-Y2
- Blue36.6 m (40 yds.)12.7 mm (½")Each38,44 $Roll of 138,44 $Supplier Number: 77010-B1
- White36.6 m (40 yds.)25.4 mm (1")Each70,01 $Roll of 170,01 $Supplier Number: 77010-W3
- Orange36.6 m (40 yds.)19.0 mm (¾")Each56,79 $Roll of 156,79 $Supplier Number: 77010-O2
- Red36.6 m (40 yds.)19.0 mm (¾")Each55,37 $Roll of 155,37 $Supplier Number: 77010-R2
- Red36.6 m (40 yds.)12.7 mm (½")Discontinued-Supplier Number: 77010-R1
- Red36.6 m (40 yds.)25.4 mm (1")Each69,17 $Roll of 169,17 $Supplier Number: 77010-R3
- White36.6 m (40 yds.)19.0 mm (¾")Each46,40 $Roll of 146,40 $Supplier Number: 77010-W2
- Blue36.6 m (40 yds.)19.0 mm (¾")Each46,37 $Roll of 146,37 $Supplier Number: 77010-B2
- Green36.6 m (40 yds.)25.4 mm (1")Each56,91 $Roll of 156,91 $Supplier Number: 77010-G3
- Green13.7 m (15 yds.)12.7 mm (½")Discontinued-Supplier Number: 77009-G1
- Supplier Number: 77009-G2
- Green13.7 m (15 yds.)25.4 mm (1")Discontinued-Supplier Number: 77009-G3
- Supplier Number: 77009-W1
- White13.7 m (15 yds.)19.0 mm (¾")Discontinued-Supplier Number: 77009-W2
- Supplier Number: 77009-W3
- Yellow13.7 m (15 yds.)12.7 mm (½")Discontinued-Supplier Number: 77009-Y1
- Supplier Number: 77009-Y2
- Supplier Number: 77009-Y3
- Supplier Number: 77011-W1
- Supplier Number: 77011-W2
- Supplier Number: 77011-W3