Disc Tab Punches, Electron Microscopy Sciences
These disc punches are produces discs (tabs) from sheet materials. Comes with adjustable side gauge for centering hole of up to 2" maximum reach.
- Maximum thickness for punching is up to 67-mil (0.17 cm)
- Ticket punching type
Ideal for punching adhesive tabs for specimen mounts or producing circle cover slips from ACLAR® films for growing cells.
: The complete unit is supplied with one punch handle and one punching die.
These disc punches are produces discs (tabs) from sheet materials. Comes with adjustable side gauge for centering hole of up to 2" maximum reach.
- Maximum thickness for punching is up to 67-mil (0.17 cm)
- Ticket punching type
Ideal for punching adhesive tabs for specimen mounts or producing circle cover slips from ACLAR® films for growing cells.
: The complete unit is supplied with one punch handle and one punching die.
Product Family Options
- DésignationHole Diameter
- : 77850-08
- : 77850-09
- : 77850-12