Staining Dish, Electron Microscopy Sciences
Staining Hellendahl Dish, 76 x 25 x 75 mm
Catalog # 100492-954
 :  70313-01
Staining Dish, Electron Microscopy Sciences
Catalog # 100492-954
Staining Dish, Electron Microscopy Sciences
Staining Dish, Electron Microscopy Sciences
Staining Hellendahl Dish, 76 x 25 x 75 mm
Price
$130.65
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Hellendahl type dish for staining or for use as a TLC developing chamber. Holds up to 8 individual 76 x 26 mm (3 x 1") slides, or 16 slides back-to-back.
These dishes are made from soda lime glass. The approximate inside dimensions L x W x D: 76 x 25 x 75 mm.
Specifications
  • Désignation:
    Staining Hellendahl Dish
  • Dimensions:
    76 x 25 x 75 mm