Horizontal Slide Tray, Electron Microscopy Sciences
Catalog # 100493-298
Supplier: Electron Microscopy Sciences
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Horizontal Tray
- Environmentally Preferable:
- Cat. no.:100493-298
- Supplier No.:71394-01
Specifications
About this item
This stainless steel tray has grooves to hold various microscope slides horizontally. Accommodates up to 60 slides.