CellSafe™ Biopsy Insert, Electron Microscopy Sciences
CellSafe™ Biopsy Insert, Electron Microscopy Sciences
CellSafe™ biopsy insert, 28×25×5 mm (1.1×1×¹/₄")
Catalog # 100501-266
Supplier #:  62327-10
CellSafe™ Biopsy Insert, Electron Microscopy Sciences
Catalog # 100501-266
CellSafe™ Biopsy Insert, Electron Microscopy Sciences
CellSafe™ Biopsy Insert, Electron Microscopy Sciences
CellSafe™ biopsy insert, 28×25×5 mm (1.1×1×¹/₄")
Price
$180.09
Log in for availability
Each Pack of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The CellSafe™ is a disposable plastic biopsy insert consisting of two hinged interlocking plastic frames with an integral nylon mesh.
  • The CellSafe™ is designed for single use only
  • Strictly avoid using in microwave processors
  • The mesh area measures 18×18×2 mm
The mesh is extra fine and therefore ensures a high level of specimen security.
The plastic frame and nylon mesh are suitable for use in common tissue processing reagents including formaldehyde solutions, Bouin's fluid, alcohol solutions, xylene, chloroform, citrus based clearing agents and paraffin wax (melting point up to 65 °C).
Specifications
  • Désignation:
    CellSafe™ biopsy insert
  • Overall Dimensions:
    28×25×5 mm (1.1×1×¹/₄")