Embryo Dishes, Electron Microscopy Sciences | Avantor
Embryo Dishes, Electron Microscopy Sciences
Embryo Dishes, Electron Microscopy Sciences
Certificates
Embryo Dishes, Electron Microscopy Sciences
Embryo Dishes, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These dishes are molded from clear glass and come with a glass lid for the viewing or staining of free-floating specimens.
Product Family Options
Informations sur le produit
  • Désignation
    Ø ext.
    Availability
    Price
  • Embryo Dish
    30 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $36.55
    $36.55
    Each
       $36.55
    Supplier Number: 70543-30
  • Embryo Dish
    45 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $58.36
    $58.36
    Each
       $58.36
    Supplier Number: 70543-45