ContactUsLinkComponent
Alcohol Burner with Ground Glass Cap, Electron Microscopy Sciences
  100498-652
Minority or Woman-Owned Business Enterprise
undefined
Alcohol Burner with Ground Glass Cap, Electron Microscopy Sciences
  100498-652
 :  64213-01
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Volume:
    100 ml (3.5 oz.)
  • Description:
    Alcohol burner
  • Cat. no.:
    100498-652
  • Supplier No.:
    64213-01
This burner offers a low heat flame, which is well suited for microscopy purposes.
  • For use with isopropyl and denatured ethyl alcohol
  • Made from soda lime glass.
Dia.× H: 75×100 mm
 : The unit is supplied with a ground glass stopper.