Dropping Bottles, Glass, Electron Microscopy Sciences
Dropping Bottles, Glass, Electron Microscopy Sciences
Dropping Bottles, Glass, Electron Microscopy Sciences
Certificates
Dropping Bottles, Glass, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These bottles deliver stains and indicator solutions drop by drop from a projecting drip tip on the side of the stopper.
A partial turn of the ground glass stopper will seal the bottle.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Clear Dropping Bottle, 50 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $252.88
    $252.88
    Log in for availability
    Pack of 4
    Pack of 4 - $176.34
    $176.34
    Each
       $252.88
    Pack of 4
       $176.34
    Supplier Number: 64220-05
  • Amber Dropping Bottle, 50 ml
    Discontinued
    Supplier Number: 64221-05
  • Amber Dropping Bottle, 100 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $316.63
    $316.63
    Log in for availability
    Pack of 4
    Pack of 4 - $230.35
    $230.35
    Each
       $316.63
    Pack of 4
       $230.35
    Supplier Number: 64221-10