MicroPoint™ Micro Dissecting Scissors, Electron Microscopy Sciences
MicroPoint™ Micro Dissecting Scissors, Electron Microscopy Sciences
MicroPoint™ Micro Dissecting Scissors, Electron Microscopy Sciences
Certificates
MicroPoint™ Micro Dissecting Scissors, Electron Microscopy Sciences
MicroPoint™ Micro Dissecting Scissors, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Scissors are made from 20 gauge stainless steel and have a 3 mm cutting edge.
Product Family Options
Product Information
  • Désignation
    Curvature
    Availability
    Price
  • Horizontal Opening
    Curved
    Discontinued
    Supplier Number: 77930-02
  • Horizontal Opening
    Straight
    Log in for availability
    Log in for availability
    Each
    Each - $959.14
    $959.14
    Each
       $959.14
    Supplier Number: 77930-01