Wood Applicators, Electron Microscopy Sciences
Product Details & Documents
These sticks are made from closed-grain Northern White Birch. This selective wood provides a smooth, splinter-free finish with good tensile strength. In addition to medical uses, these sticks are ideal for spreading ointments and salves, stirring, and mixing.
Specifications
- Désignation:Double Bevel Applicator Stick
- Dimensions:7 x 5/32"