White Porcelain Plate, Electron Microscopy Sciences
Twelve cavities on a white porcelain plate. Used for staining and color reactions. They measure L x W: 118 x 90 mm (4 5/8 x 3 1/2"). Cavity depth: 6.4 mm (1/4").
Twelve cavities on a white porcelain plate. Used for staining and color reactions. They measure L x W: 118 x 90 mm (4 5/8 x 3 1/2"). Cavity depth: 6.4 mm (1/4").
Product Family Options
- Désignation
- : 71562-01
- White Porcelain PlateEach$1,088.78Pack of 12$788.86: 71562-06