V-Vials®; Clear With Screw Cap, Electron Microscopy Sciences
V-Vials®; Clear With Screw Cap, Electron Microscopy Sciences
Certificates
V-Vials®; Clear With Screw Cap, Electron Microscopy Sciences
V-Vials®; Clear With Screw Cap, Electron Microscopy Sciences
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
V-Vials are ideal for: micro mixing, homogenization, centrifugation, concentration, extraction, derivatization, radiosynthesis, hybridization, anaerobic reactions, and catalytic hydrogenation.
  • All components can be autoclaved
  • Graduated and clear
These vials are made from Wheaton-33 low extractable borosilicate glass and they safeguard against any change of product pH caused by alkali, which can be extracted from ordinary glass. Wheaton V-Vials are exceptionally sturdy vials with a conical interior bottom. Their special features insure maximum security and economy in handling, storing and shipping of costly micro and semi-micro samples.
Screw cap has solid top and TFE-faced has white (styrene butadiene) rubber liner.
 : They come packed 12 vials in a hinged lid storage box.
Product Family Options
Informations sur le produit
  • Capacité
    Avec bouchon
    Availability
    Price
  • 0.30 ml
    Oui
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $473.47
    $473.47
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $332.59
    $332.59
    Each
       $473.47
    Pack of 12
       $332.59
    Supplier Number: 72647-01
  • 1.00 ml
    Oui
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $534.74
    $534.74
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $385.20
    $385.20
    Each
       $534.74
    Pack of 12
       $385.20
    Supplier Number: 72647-02
  • 3.00 ml
    Oui
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $370.52
    $370.52
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $519.60
    $519.60
    Each
       $370.52
    Pack of 12
       $519.60
    Supplier Number: 72647-07
  • 5.00 ml
    Oui
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $724.07
    $724.07
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $505.95
    $505.95
    Each
       $724.07
    Pack of 12
       $505.95
    Supplier Number: 72647-08
  • 2.00 ml
    Oui
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $646.03
    $646.03
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $463.26
    $463.26
    Each
       $646.03
    Pack of 12
       $463.26
    Supplier Number: 72647-05