À propos de cet article
For the orderly visual display and safe transport of microscope slides.
This loose-leaf binder has ten vinyl View-Pack pages (hold 160 slides). Each page holds 16 slides 75 x 25 mm (3 x 1") in individual pockets with a white back and clear front. A flap in the center keeps slides from falling out. Simply fold the page at the center crease to retrieve slides. The 22 x 27 cm (8 1/2 x 10 1/2") View-Pack pages comes with a standard 23 x 30 cm (9 x 12") 3-ring binder.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.