Shell Vials, Electron Microscopy Sciences
À propos de cet article
These vials are optically clear with straight-sides. Made from Type1, N-51A borosilicate glass. Fixation, dehydration, and infiltration procedures can be done in the same vial. The larger size vials can be used as an embedding media mixture container or for sample storage.
Delivery information: The vials come with polyethylene open bottom snap-caps, Titeseal®, which are packed separately. Except the 4- dram size vial which comes without a closure. We recommend the use of our cork stoppers.
6 Options Available Below
Product Details & Documents
These vials are optically clear with straight-sides. Made from Type1, N-51A borosilicate glass. Fixation, dehydration, and infiltration procedures can be done in the same vial. The larger size vials can be used as an embedding media mixture container or for sample storage.
Delivery information: The vials come with polyethylene open bottom snap-caps, Titeseal®, which are packed separately. Except the 4- dram size vial which comes without a closure. We recommend the use of our cork stoppers.
Product Family Options
Informations sur le produit
- DésignationAvec bouchonAvailabilityPrice
- Shell vials, 1 DramOuiEach$179.67Pack of 144$136.56Supplier Number: 72631-10
- Shell vials, 2 DramOuiEach$342.03Pack of 144$260.35Supplier Number: 72631-20
- Shell vials, 3 DramOuiEach$227.91Pack of 144$191.62Supplier Number: 72631-30
- Shell vials, 0.5 DramOuiEach$176.49Pack of 144$152.88Supplier Number: 72630-05
- Shell vials, 4 DramNonEach$132.89Pack of 144$101.28Supplier Number: 72631-40
- Shell vials, 7 DramOuiEach$500.41Pack of 144$381.89Supplier Number: 72631-70