Scintillation Vials, Electron Microscopy Sciences
Scintillation Vials, Electron Microscopy Sciences
Certificates
Scintillation Vials, Electron Microscopy Sciences
Scintillation Vials, Electron Microscopy Sciences
6 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Vials are available in Wheaton 180® low potassium borosilicate glass, High Density Polyethylene (HDPE) and Polyethylene Terephthalate (PET).
  • Screw caps size 22-400 is made from Urea with Poly-seal cone liner
  • Vial measures 28 mm diameter × 57 mm height (with cap off), and 61 mm height (with cap on)
Opaque, high-density polyethylene vials are economically priced. They are 20 ml in capacity and they measure 27 mm diameter × 61 mm height (with cap). The screw caps are 22-400 and they are made from polypropylene with a foam- polyethylene liner.
For optimal performance in the laboratory, High Density Polyethylene (HDPE) vials provide a combination of features previously unavailable in a counting vial. Safe disposal is accomplished by incineration. No harmful gas is generated (only CO₂ and H₂O are created). These vials are lightweight, virtually unbreakable and they have the clarity of glass. PET vials also offer low permeability to solvents and minimal background count. These vials hold 20 ml of fluid and they measure 27 mm diameter, 61 mm height (with cap on). The screw caps are 22-400 and are made from polypropylene with a foamed polyethylene liner.
 : Wheaton '180'® low potassium borosilicate glass meets ASTM Type I Class A and USP Type I standards.
 : Wheaton glass and polyethylene vials are packed in individual cells in trays of 100. Vials in trays are shrunk-wrapped in plastic, 5 trays to a case. Caps are packed separately in polyethylene bags of 100. PET vials and caps are packaged separately.
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Matière
    Avec bouchon
    Availability
    Price
  • Scintillation Vial
    20.00 ml
    HDPE
    Oui
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $174.54
    $174.54
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $133.24
    $133.24
    Each
       $174.54
    Pack of 100
       $133.24
    Supplier Number: 72632-PE
  • Scintillation Vial
    20.00 ml
    HDPE
    Oui
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $739.06
    $739.06
    Log in pour la disponibilité
    Pack of 500
    Pack of 500 - $627.18
    $627.18
    Each
       $739.06
    Pack of 500
       $627.18
    Supplier Number: 72634-PE
  • Scintillation Vial
    20.00 ml
    Polyethylene Terephthalate
    Oui
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $214.39
    $214.39
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $163.10
    $163.10
    Each
       $214.39
    Pack of 100
       $163.10
    Supplier Number: 72632-PT
  • Scintillation Vial
    20.00 ml
    Polyethylene Terephthalate
    Oui
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $957.76
    $957.76
    Log in pour la disponibilité
    Pack of 500
    Pack of 500 - $704.32
    $704.32
    Each
       $957.76
    Pack of 500
       $704.32
    Supplier Number: 72634-PT
  • Scintillation Vial
    20.00 ml
    Wheaton 180® glass
    Oui
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 100
    Box of 100 - $294.38
    $294.38
    Log in pour la disponibilité
    Each
    Each - $294.38
    $294.38
    Box of 100
       $294.38
    Each
       $294.38
    Supplier Number: 72632
  • Scintillation Vial
    20.00 ml
    Wheaton 180® glass
    Oui
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 500
    Box of 500 - $1,518.90
    $1,518.90
    Log in pour la disponibilité
    Each
    Each - $1,518.90
    $1,518.90
    Box of 500
       $1,518.90
    Each
       $1,518.90
    Supplier Number: 72634