Vial Rack, Electron Microscopy Sciences
Vial Rack, Electron Microscopy Sciences
Vial Rack, Electron Microscopy Sciences
Certificates
Vial Rack, Electron Microscopy Sciences
Vial Rack, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This rugged polypropylene rack holds 48 vials, size 1 dram (15 x 45 mm); or 4 ml tissue culture vials; or 2 mL serum vials in 4 x 12 configuration with alpha-numeric indexing.
The rack is constructed from chemically resistant, autoclavable (121 °C for 20 minutes) polypropylene and features corner posts to assure stability when stacking empty.
Product Family Options
Product Information
  • Désignation
    Dimensions
    Availability
    Price
  • Rack for 1 Dram Shell Vial
    94 x 266 x 28 mm (3 3/4 x 10 5/8 x 1 1/8")
    Log in for availability
    Log in for availability
    Each
    Each - $150.22
    $150.22
    Each
       $150.22
    Supplier Number: 72641-01
  • Rack for 1 Dram Shell Vial
    94 x 266 x 28 mm (3 3/4 x 10 5/8 x 1 1/8")
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 5
    Case of 5 - $537.48
    $537.48
    Log in for availability
    Each
    Each - $537.48
    $537.48
    Case of 5
       $537.48
    Each
       $537.48
    Supplier Number: 72641-05