Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Vertical Staining Jar with Glass Lid
- Cat. no.:100490-752
- Supplier No.:70318-04
Specifications
About this item
An all glass rectangular staining jar which accommodates 8 to 16 slides and comes with a glass lid.
Its wide top eliminates the need for forceps in the handling of the slides. It accommodates slides up to 103×39 mm (4×1½") in size.