Vacuum Desiccators, Electron Microscopy Sciences
Vacuum Desiccators, Electron Microscopy Sciences
Vacuum Desiccators, Electron Microscopy Sciences
Certificates
Vacuum Desiccators, Electron Microscopy Sciences
Vacuum Desiccators, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All desiccators are made of clear polycarbonate.
  • Large interior volume
  • Extra heavy walls
  • Stopcock with a "PTFE" plug
  • Ribbed Knob
  • Neoprene "O" ring
  • Fully visible
Accessories information: Vacuum Jars are replacement jars for all Dry-Seal desiccators. The covers, without sleeve valves and the sleeve valves are replacements for all Dry Seal® vacuum desiccators. Retaining rings are for sleeve valves. Sealing rings are molded from chemically resistant Viton® rubber.
Product Family Options
Informations sur le produit
  • Désignation
    Overall Height
    Inner Diameter
    Flange O.D.
    Clearance Above Plate
    Plate Size
    Availability
    Price
  • Medium Vacuum Desiccator
    260 mm
    197 mm
    230 mm
    157 mm
    190 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $438.55
    $438.55
    Each
       $438.55
    Supplier Number: 71232
  • Large Vacuum Desiccator
    311 mm
    240 mm
    273 mm
    197 mm
    230 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $595.45
    $595.45
    Each
       $595.45
    Supplier Number: 71234
  • Small Vacuum Desiccator
    206 mm
    149 mm
    71 mm
    121 mm
    140 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $344.57
    $344.57
    Each
       $344.57
    Supplier Number: 71230
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Aluminum Plates, 250 mm
    Aluminum Plates, 250 mm
    102097-462
    Log in pour la disponibilité
    Each - $426.87
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Vacuum Jars, 250 mm
    Vacuum Jars, 250 mm
    100493-238
    Log in pour la disponibilité
    Each - $805.27
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Covers without Sleeve Valves, 170 mm, 40/35
    Covers without Sleeve Valves, 170 mm, 40/35
    100493-242
    Log in pour la disponibilité
    Each - $347.26
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Sealing Rings, 100 mm
    Sealing Rings, 100 mm
    100493-258
    Log in pour la disponibilité
    Each - $195.86