JEOL Strip Apertures, Electron Microscopy Sciences
À propos de cet article
Multihole strip aperture made from Platinum-Iridium (95:5).
8 Options Available Below
Product Details & Documents
Multihole strip aperture made from Platinum-Iridium (95:5).
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 49210-01
- Supplier Number: 49210-02
- Supplier Number: 49210-03
- JEOL, Hole Sizes: 100, 200, 300 µm; 9.0×2.35×0.10 mmDiscontinued-Supplier Number: 49210-04
- Strip Apertures, Hole Sizes: 20, 60, 100 µm; 10.0×2.5×0.10 mmLog in pour la disponibilitéEach$1,751.14Supplier Number: 49213-01
- Strip Apertures, Hole Sizes 20, 30, 30, 50 µm; 31.0×5.0×0.10 mmDiscontinued-Supplier Number: 49214-01
- Supplier Number: 49211-01
- Supplier Number: 49211-02
- Supplier Number: 49211-03
- Hitachi, Hole Sizes: 30, 70, 100, 200 µm; 45.0×4.0×0.10 mm; 45.0×4.0×0.10 mmDiscontinued-Supplier Number: 49212-02
- Supplier Number: 49212-03