About this item
Precision machined from aluminum
These special chucks are made to accommodate small specimen blocks, which have been polymerized in Eppendorf tubes. In addition, we also have chucks that hold all sizes of flat embedding blocks (fitting all Leica and RMC instruments), and chucks for LKB instruments.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.