Tri-Corn Beakers, Plastic, Disposable, Electron Microscopy Sciences
Tri-Corn Beakers, Plastic, Disposable, Electron Microscopy Sciences
Tri-Corn Beakers, Plastic, Disposable, Electron Microscopy Sciences
Tri-Corn Beakers, Plastic, Disposable, Electron Microscopy Sciences
Autoclavable beakers that can withstand temperatures up to 121 °C.
  • Metric graduations
  • Three dripless pouring spouts
  • Resistant to most chemicals
Product Family Options
  • Capacité
    Hauteur
    Ø ext.
    Intervalle de graduation
  • 100 ml (3.4 oz.)
    73 mm
    53 mm
    10 ml
    Each
    Each - $134.24
    $134.24
    Pack of 100
    Pack of 100 - $120.05
    $120.05
    Each
       $134.24
    Pack of 100
       $120.05
     : 60970
  • 250 ml
    90 mm
    72 mm
    10 ml
    Each
    Each - $206.01
    $206.01
    Pack of 100
    Pack of 100 - $145.08
    $145.08
    Each
       $206.01
    Pack of 100
       $145.08
     : 60972
  • 400 ml
    110 mm
    80 mm
    20 ml
    Each
    Each - $247.21
    $247.21
    Pack of 100
    Pack of 100 - $194.13
    $194.13
    Each
       $247.21
    Pack of 100
       $194.13
     : 60974
  • 800 ml
    134 mm
    107 mm
    50 ml
    Each
    Each - $374.30
    $374.30
    Pack of 100
    Pack of 100 - $223.49
    $223.49
    Each
       $374.30
    Pack of 100
       $223.49
     : 60976
  • 1000 ml
    147 mm
    112 mm
    50 ml
    Each
    Each - $499.29
    $499.29
    Pack of 100
    Pack of 100 - $284.05
    $284.05
    Each
       $499.29
    Pack of 100
       $284.05
     : 60978