Non-Marking Heel Grounder, Electron Microscopy Sciences
À propos de cet article
E.Z. Close is your low cost logical choice heel grounder.
- The Velcro® fastening system is adjust-able and comfortable
- Charge decay: less than 0.01 sec.; 2.4×108 Ω
They are made from premium grade non-marking conductive rubber that is highly abrasion and wear resistant.
Certifications: Meets DOD-263A standards (USA).
Delivery information: Includes: 18" conductive tab and 1¹/₂", two-layer, rubber cup. Built in resistor.
1 Options Available Below
Product Details & Documents
E.Z. Close is your low cost logical choice heel grounder.
- The Velcro® fastening system is adjust-able and comfortable
- Charge decay: less than 0.01 sec.; 2.4×108 Ω
They are made from premium grade non-marking conductive rubber that is highly abrasion and wear resistant.
Certifications: Meets DOD-263A standards (USA).
Delivery information: Includes: 18" conductive tab and 1¹/₂", two-layer, rubber cup. Built in resistor.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Heel Grounder with 1 Mega Ohm Resistor BlueDiscontinued-Supplier Number: 60619-10
- Supplier Number: 60619-30