Tray, Dispensing, high density polyethylene, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
These trays, which are ¹/₄" thick, are fabricated from either low or high density polyethylene.
- Tray is suitable for specimen preparation procedures
They come complete with a polyethylene dispensing valve which is located as close to the bottom as possible on the corner.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together