Transfer Pipette, Polypropylene, Electron Microscopy Sciences
Transfer Pipette, Polypropylene, Electron Microscopy Sciences
Certificates
Transfer Pipette, Polypropylene, Electron Microscopy Sciences
Transfer Pipette, Polypropylene, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Transfer pipette that works as a syringe for liquid transfer. Each pipette has calibration markings from 0 to 12 ml in 0.2 ml increments. Tubing may be attached to the tip to suit your application, and tips may also be cut to facilitate aspiration and delivery of fluids of varying viscosities.
  • Inexpensive and disposable
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Transfer Pipettes, Straight, 12 mL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 50
    Box of 50 - $160.27
    $160.27
    Log in pour la disponibilité
    Each
    Each - $160.27
    $160.27
    Box of 50
       $160.27
    Each
       $160.27
    Supplier Number: 70986-00
  • Transfer Pipettes, Curved, 12 mL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 50
    Box of 50 - $195.51
    $195.51
    Log in pour la disponibilité
    Each
    Each - $195.51
    $195.51
    Box of 50
       $195.51
    Each
       $195.51
    Supplier Number: 70986-20
  • Transfer Pipettes, Straight, 12 mL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $978.51
    $978.51
    Pack of 50
       $978.51
    Supplier Number: 70986-00