Tooled Neck Vials, Electron Microscopy Sciences
À propos de cet article
These vials are made from Type 1, Class B borosilicate glass. Their tooled necks ensure accurate fit with 'snap-in' closures. They are ideal for storage of specimens and sample blocks.
Delivery information: The 1 and 2 dram sizes come with an open-bottom closure and all of the other sizes come with a closed bottom, two-piece closure.
Packaging: Closures are packed separately.
5 Options Available Below
Product Details & Documents
These vials are made from Type 1, Class B borosilicate glass. Their tooled necks ensure accurate fit with 'snap-in' closures. They are ideal for storage of specimens and sample blocks.
Delivery information: The 1 and 2 dram sizes come with an open-bottom closure and all of the other sizes come with a closed bottom, two-piece closure.
Packaging: Closures are packed separately.
Product Family Options
Informations sur le produit
- DésignationAvec bouchonAvailabilityPrice
- Tooled Neck Vials, 3DROuiEach$327.13Pack of 144$249.70Supplier Number: 72646-03
- Tooled Neck Vials, 1DROuiEach$235.97Pack of 144$180.29Supplier Number: 72646-01
- Tooled Neck Vials, 5DROuiEach$518.33Pack of 144$394.95Supplier Number: 72646-05
- Tooled Neck Vials, 4DROuiEach$391.18Pack of 144$298.67Supplier Number: 72646-04
- Tooled Neck Vials, 2DROuiEach$218.94Pack of 144$166.88Supplier Number: 72646-02